Troemner 318ST Economical S.S. Class 7 Weight Set; 500 mg to 1 mg with Traceable Certificate
Representative image only.
Troemner Mfr # 318ST – Item # EW-01029-73
Make the most of your balance—calibrate regularly!
  • Adjusted to ANSI/ASTM E617 Class 7
PRODUCT OPTIONS
$305.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass 7
  • Mass CertificationNIST
  • Height (in)0.875
  • Width (in)1.8906
  • Length (in)4.5156
  • Contents of Set500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg, 5mg, (2) 2mg, 1mg; polycarbonate case
  • DescriptionEconomical S.S. Class 7 Weight Set; 500 mg to 1 mg with Traceable Certificate
Includes
Hinged polypropylene box with snap-tight closure and NIST-traceable calibration certificate.
Key Features
  • Adjusted to ANSI/ASTM E617 Class 7
More About this Item
This economical set is ideal for general-purpose applications. Weight sets are designed to achieve any total mass by combining smaller masses. All weighs feaature a satin finish surface and are marked with their nominal mass value.