Troemner 409 Heavy-Duty, Variable Speed Mixer, 120 VAC
Representative image only.
Troemner Mfr # 409 – Item # EW-04340-10
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Speed (rpm)50
  • Max Speed (rpm)500
  • ChuckCollet
  • Max Torque (in-oz)400
  • Motor hp1/4
  • DescriptionHeavy-Duty, Variable Speed Mixer, 120 VAC
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Heavy-duty, variable-speed mixer features a 1/4-hp motor for mixing any fluid with a viscosity up to 150,000 cp. This mixer includes a heavy-duty double clamp.