TSI Alnor AXD620 Micromanometer System with Data Logging Software Kit
Representative image only.
TSI Mfr # AXD620 – Item # EW-10375-37
Premium features make it ideal for HVAC, environmental safeguards, commissioning, process control and system balancing
  • Measure differential and static pressure from -15 to +15 in. H2O (-3735 to +3735 Pa)
  • Calculates and displays velocity when using a Pitot tube
  • Calculates volumetric flow rate in duct from velocity and user-input duct size and shape
  • Records data points in duct traverse using sampling function
  • Data logging with time and date stamp
$1,010.00USD / Each
sign in for your price.
$146.00 - $216.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Air Velocity (ft/min)15500
  • Min Air Velocity (ft/min)250
  • Min Temperature (° F)40
  • Max Temperature (° F)113
  • Min Temperature (° C)5
  • Max Temperature (° C)45
  • Data LoggingYes
  • Data Storage (samples)12,700 samples and 100 test IDs
  • Logging Intervals1 second to 1 hour
  • Width (in)3 5/16
  • Height (in)1 3/4
  • Length (in)7
  • DescriptionMicromanometer System with Data Logging Software Kit
Conformite Europeenne
Includes
Meter, hard-sided carrying case, 4 AA batteries, two 1.2m long rubber tubes, static pressure tip, LogDat2 downloading software and USB cable, manufacturer provided calibration certificate.
Key Features
  • Measure differential and static pressure from -15 to +15 in. H2O (-3735 to +3735 Pa)
  • Calculates and displays velocity when using a Pitot tube
  • Calculates volumetric flow rate in duct from velocity and user-input duct size and shape
  • Records data points in duct traverse using sampling function
  • Data logging with time and date stamp
  • Programmable K factors
More About this Item
The Alnor® AXD620 Micromanometer is a rugged, compact, comprehensive micromanometer that measures pressure. It can be used with pitot tubes to measure velocity and calculate flow rates with user–input duct size and shape.
Ideal applications include HVAC commissioning and troubleshooting, testing and balancing, Pitot tube duct traverses, static pressure measurements and differential pressure measurements.