TSI AVM410 Thermoanemometer Meter Kit

Representative image only.
TSI Mfr # AVM410 – Item # EW-10508-73
Troubleshoot HVAC systems easily
  • Accurate air velocity measurement
  • Integrated probe attachment
  • Large, easy-to-read display

$840.00USD / Each
sign in for your price.

$306.00 - $379.00USD / Each

for an additional
$306.00USD / Each
for an additional
$379.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Air Velocity (ft/min)4000
  • Min Air Velocity (ft/min)0
  • Air Velocity Resolution1 ft/min (0.01 m/s)
  • Air Velocity Accuracy±5% of reading or ±5 ft/min (±0.025 m/s) (whichever is greater)
  • Number Of Temperature Channels1
  • Min Temperature (° F)0
  • Max Temperature (° F)200
  • Min Temperature (° C)-18
  • Max Temperature (° C)93
  • Temperature Accuracy±0.5°F (0.3°C)
  • Temperature Resolution0.1°F (0.1°C)
  • Data LoggingNo
  • Probe Diameter (in)9/32
  • Battery4 AA (included)
  • DescriptionThermoanemometer Meter Kit
Two year warranty

Key Features

  • Accurate air velocity measurement
  • Integrated probe attachment
  • Large, easy-to-read display

More About this Item

These thermal anemometers are a solid choice for a digital air velocity meter without compromising accuracy or precision. They are excellent for troublehshooting HVAC systems and conducting commissioning work.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat