Cole-Parmer Tube Rotator Microcentrifuge Tube Holder Disc, 46 Tube Capacity
Representative image only.
Cole-Parmer Item # EW-00077-11
Quickly and easily replace tube holder without tools
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers an end over end action
  • Holds 46 x 1.5 mL tubes
  • 1 year warranty
  • Previous part number SB3/1
PRODUCT OPTIONS
$226.75USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Number of Tubes46
  • Tube Size10 to 11.5 mm dia
  • DescriptionTube Rotator Microcentrifuge Tube Holder Disc, 46 tube capacity
  • Warranty1 Year
1 Year Warranty
Key Features
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers an end over end action
  • Holds 46 x 1.5 mL tubes
  • 1 year warranty
  • Previous part number SB3/1