U E Systems DHC-2 Deluxe Noise Isolating Headset
Representative image only.
U E Systems Mfr # DHC-2 – Item # EW-98853-35
Blocks out intense sounds
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUe Systems Deluxe Noise Isolating Headset
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This heavy duty headset is designed to block out intense sounds (over 23 dB of noise attenuation) often found in industrial environments so the user may easily hear the sounds received by the Ultraprobe 9000.