U E Systems UE 201 Grease Caddy Ultrasonic Lube Assessment Instrument
Representative image only.
U E Systems Mfr # UE 201 – Item # EW-59501-46
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGrease Caddy Ultrasonic Lube Assessment Instrument
  • Warranty5 Years
5 Year Warranty
Key Features
  • 5 year warranty
More About this Item
One of the most common causes of bearing failure is over lubrication. The Grease Caddy prevents over lubrication and helps prolong bearing life. Using advanced ultrasonic technology, the Grease Caddy helps you isolate and clearly hear bearing sounds in most noisy plant environments. It is so sensitive you will hear when grease enters a bearing and recognize when to stop applying your lubrication. The Grease Caddy easily attaches to most common grease guns.
FEATURES
  • Swivel base to adjust for odd angles
  • A mini-spot light helps users see their lube connections in low light situations
  • Sensitivity Adjust Dial
  • LED Intensity display
  • Comfortable headphones