U E Systems UP100KT Ue Systems Comprehensive Ultrasonic Inspection Kit
Representative image only.
U E Systems Mfr # UP100KT – Item # EW-98853-34
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUe Systems Comprehensive Ultrasonic Inspection Kit
  • Warranty5 Years
5 Year Warranty
Key Features
  • 5 year warranty
More About this Item
Comprehensive Kit
Includes: Pistol housing with LED bargraph meter, 8 position sensitivity selection, low battery indicator, scanning module, rubber focusing probe, stethoscope module, warble tone generator(WTG-1) with battery charger, headset, 9 volt alkaline battery (replaceable), carrying case and comprehensive instruction manual
APPLICATIONS
  • Pressure and Vacuum Lead Detection
  • Valve seat lead detection
  • Exhaust System Leaks
  • Heat Exchangers Boilers Condensers
  • Steam Trap Inspection
  • Bearing Testing
  • Gear / Gear Box Inspection
  • Wind Noise / Water Leaks
  • General Mechanical Inspection and Trouble Shooting
  • Tanks, Pipes, etc. Leak Testing
  • Electrical Inspection