Unico Spectrophotometer Temperature Control System, 115 VAC
Representative image only.
Cole-Parmer Mfr # S-2150-107P – Item # EW-83057-66
Customize your spectrophotometer
  • Compatible with Unico visible spectrophotometers
  • Easy to install and connect
  • Ensure accuracy and precision
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With83059-10
  • Power (VAC)115
  • DescriptionSpectrophotometer Temperature Control System, 115 VAC
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with Unico visible spectrophotometers
  • Easy to install and connect
  • Ensure accuracy and precision
  • 1 year warranty
More About this Item
Customize your Unico spectrophotometer with cell holders, sample chambers, and other accessories.
Temperature Control System for Unico Spectrophotometers. With external controller and Peltier 10 mm cell holder. 115 VAC, 15 to 40°C.