United Electric 2W2D00P16 Discrete Input Pressure Switch, 500 psi, 40mA@12-30VDC

Representative image only.
United Electric Mfr # 1XSWLLP16 – Item # EW-68457-09
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±0.5% of adjustable range
  • Min Pressure (PSI)0
  • Max Pressure (PSI)500
  • Casealuminum
  • Process Connection1/4\" NPT(F)
  • Battery TypeDiscrete input
  • Electrical Connections1/2\" NPT(F) conduit
  • Electrical terminationScrew terminals
  • Output12 to 30 VDC
  • Enclosure RatingNEMA 4X, IP66
  • DescriptionDiscrete Input Pressure Switch, 500 psi, 40mA@12-30VDC
  • Warranty3 year
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

FEATURES

  • Legendary UE reliability
  • Hazardous location
  • Class I, Div 2; Class II, Div 2; Class III
  • Worldwide approvals and certifications
  • Large process display
  • 100% programmable set-point and deadband
  • Terminal block wiring
  • NEMA 4X / IP 66 die cast aluminum enclosure, epoxy powder coated and gasketed


One series

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat