United Electric 4W3A01P16 Switch 0/500psi 90-130VAC Power Input 24-280 VAC

Representative image only.
United Electric Mfr # 1XSWHHP16 – Item # EW-68457-21
  • 3 year warranty

PRODUCT OPTIONS

$1,383.00USD / Each

$152.00 - $225.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±0.5% of adjustable range
  • Min Pressure (PSI)0
  • Max Pressure (PSI)500
  • Casealuminum
  • Process Connection1/4\" NPT(F)
  • Electrical Connections1/2\" NPT(F) conduit
  • Electrical terminationScrew terminals
  • Output24 to 280 VAC
  • Enclosure RatingNEMA 4X, IP66
  • DescriptionSwitch 0/500psi 90-130VAC Power Input 24-280 VAC
  • Warranty3 year
  • Power90 to 130 VAC
3 Year Warranty

Key Features

  • 3 year warranty

More About this Item

FEATURES

  • Legendary UE reliability
  • Hazardous location
  • Class I, Div 2; Class II, Div 2; Class III
  • Worldwide approvals and certifications
  • Large process display
  • 100% programmable set-point and deadband
  • Terminal block wiring
  • NEMA 4X / IP 66 die cast aluminum enclosure, epoxy powder coated and gasketed


One series

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat