United Electric E121-8BC Explosion-Proof Temperature Switch, 350/640F; Remote Mount
Representative image only.
United Electric Mfr # E121-8BS – Item # EW-39008-30
  • 2 year warranty
PRODUCT OPTIONS
$864.50USD / Each
sign in for your price.
$209.00 - $279.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • RelaysSPDT
  • Min Temperature (° F)350
  • Max Temperature (° F)640
  • DescriptionExplosion-Proof Temperature Switch, 350/640F; Remote Mount
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Legendary UE reliability
  • Explosion-proof
  • Class I, Division 1 & 2 (Zone 1); Class II, Div. 1 and 2; Class III
  • Worldwide approvals and certifications
  • Terminal block wiring
  • Single conduit
  • Enclosure design permits safe and easy access for wiring
  • External adjustment via reference dial
  • Die cast aluminum enclosure, epoxy powder coated and gasketed