Uniweld 53650 Electronic Refrigerant Charging Scale, +/-0.2 oz/ 10 g or +/-0.5oz/20 g at 176 lbs
Representative image only.
Uniweld Mfr # 53650 – Item # EW-07164-63
Battery powered charging scale
  • LCD displays either pounds/ounces or Kilograms/Grams
  • Accuracy of ±0.2 oz./10g at up to 110 lbs
  • Accuracy of ±0.5 oz/20g on loads up to 176 lbs
  • Battery powered
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectronic Refrigerant Charging Scale, +/-0.2 oz/ 10 g or +/-0.5oz/20 g at 176 lbs
  • Warranty1 year
1 Year Warranty
Key Features
  • LCD displays either pounds/ounces or Kilograms/Grams
  • Accuracy of ±0.2 oz./10g at up to 110 lbs
  • Accuracy of ±0.5 oz/20g on loads up to 176 lbs
  • Battery powered
  • 1 year warranty
More About this Item
Optional charging module (model 07164-64) converts scale into an automated charging system and compatible with all refrigerants including R-410A as well as 120 VAC adapter (model 07164-64) are sold separately.