Uvex by Honeywell 11150350 Safety Glasses, Clear Lens, Black Frame
Representative image only.
Honeywell Mfr # 11150350 – Item # EW-81653-20
Available for sale in the US only
  • Enjoy a nonslip fit without pressure on temples or ears
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorClear
  • ComplianceANSI Z87.1-2010; CSA Z94.3
  • DescriptionSafety Glasses, Clear Lens, Black Frame
  • Warranty1 year
1 Year WarrantyCanadian Standards Association
Key Features
  • Enjoy a nonslip fit without pressure on temples or ears
  • 1 year warranty
More About this Item
Unitary curved polycarbonate lens with Parallel Vision Control™ achieves 180° clarity of view with no distortion. Frames are lightweight and balanced—a virtually weightless feel for hours of comfort. Include an eyewear retainer cord. Clear and silver inside/outside (I/O) lenses provide 100% UV protection.
Compliance: ANSI Z87.1-2010, CSA Z94.3