Uvex by Honeywell Bionic Face Shield, Clear, uncoated
Representative image only.
Uvex by Honeywell Mfr # S8500 – Item # EW-81690-11
Available for sale in the US only
  • Full-shield design provides built-in chin and top-of-head protection
  • Lightweight design, foam backing, and customizable fit make this face shield ideal for extended use
  • Fits comfortably over most goggles and respirators
  • Compliance: ANSI Z87.1-2003, CSA Z94.3
$52.50USD / Each
BUY MORE & SAVE
  • 1-9$52.50USD / Each
  • 10+$47.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorClear
  • Lens CoatingUncoated
  • ComplianceANSI Z87.1-2003; CSA Z94.3
  • DescriptionFace Shield, Clear, uncoated
Key Features
  • Full-shield design provides built-in chin and top-of-head protection
  • Lightweight design, foam backing, and customizable fit make this face shield ideal for extended use
  • Fits comfortably over most goggles and respirators
  • Compliance: ANSI Z87.1-2003, CSA Z94.3
More About this Item
Shield provides secondary protection for workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Full shield design provides built-in chin and top-of-head protection. Lightweight design, foam backing, and customizable fit make this face shield ideal for extended use. Fits comfortably over most goggles and respirators.
Compliance: ANSI Z87.1-2003 and CSA Z94.3