Uvex by Honeywell S0600 Safety Eyewear, Clear lens
Representative image only.
Uvex by Honeywell Mfr # S0600 – Item # EW-86497-60
Available for sale in the US only
  • Range of comfort and adjustability features for all-day wear
$29.00USD / Each
BUY MORE & SAVE
  • 1-9$29.00USD / Each
  • 10+$26.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorClear
  • Lens CoatingUltra-dura® (hard coating)
  • ComplianceANSI Z87.1-2010; CSA Z94.3 (with cushioning only)
  • DescriptionSafety Eyewear, Clear lens
Canadian Standards Association
Key Features
  • Range of comfort and adjustability features for all-day wear
More About this Item
Flexible design allows you to wear these as safety glasses, or attach the flame-resistant headband to wear as goggles. Also, leave the face cushioning inner frame on for extra comfort during extended-wear applications, remove it for a standard safety glasses feel. The cushioning seals your eyes from dirty, high-particulate work environments such as grinding or sanding. Wraparound style enhances peripheral vision and eye coverage. Top and bottom venting helps minimize fogging.
Compliance: ANSI Z87.1-2010 (with or without cushioning), CSA Z94.3 (with cushioning only)