Uvex by Honeywell S0620HS Seismic Safety Glasses, Blue Frame, Clear, HydroShield Antifog Lens; 10/BX
Representative image only.
Uvex by Honeywell Mfr # S0620HS – Item # EW-86498-02
Protects you in any environment
  • Protects against impact from sun, wind and dust, as well as airborne debris All-day comfort with its cushioning foam, soft and flexible nasal pads, and padded temple tips High-quality adjustable and flame-resistant headband High-performance lens coatings combined with top and bottom indirect ventilation minimize fogging for clearer, safer vision Exceptional peripheral vision and coverage with wrap-around 8-base dual-lens design
  • Metallic Blue Frame
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlue
  • Lens ColorClear
  • Lens CoatingAntiscratch, Antifog
  • ComplianceANSI 787.1+, CSAZ94.3
  • DescriptionSafety Glasses, Blue Frame, Clear, HydroShield Antifog Lens; 10/BX
Key Features
  • Protects against impact from sun, wind and dust, as well as airborne debris All-day comfort with its cushioning foam, soft and flexible nasal pads, and padded temple tips High-quality adjustable and flame-resistant headband High-performance lens coatings combined with top and bottom indirect ventilation minimize fogging for clearer, safer vision Exceptional peripheral vision and coverage with wrap-around 8-base dual-lens design
  • Metallic Blue Frame
More About this Item
Polycarbonate lens is clear and features an antiscratch and HydroShield antifog hard coating. The glasses come with removable temples, headband and cushioned subframe. Additional coatings, as well as reading magnifiers in three strengths is also available.
Compliance: Certified to meet ANSI 787.1+, CSAZ94.3