Uvex by Honeywell S2530C TG Safety Glasses, Clear Lens, Patriot Frame
Representative image only.
Uvex by Honeywell Mfr # S2530C – Item # EW-86322-30
Available for sale in the US only
  • A comfortable fit for virtually any size face
  • Adjustable temple length and lens inclination
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorRed/White/Blue
  • Temple StyleDuoflex®
  • Lens ColorClear
  • Lens CoatingUvextreme® (reduces fogging, scratches, and static)
  • ComplianceANSI Z87.1-2010; CSA Z94.3; military V0
  • DescriptionTG Safety Glasses, Clear Lens, Patriot Frame
Key Features
  • A comfortable fit for virtually any size face
  • Adjustable temple length and lens inclination
More About this Item
Features an anti-fog Uvextreme® coating with additional anti-scratch and anti-static properties. An economical alternative to expensive prescription safety glasses—eyewear fits securely over the glasses (OTG).
Compliance: ANSI Z87.1-2010, CSA Z94.3, military V0