Uvex by Honeywell S3500 afety Glasses, Silver/Navy frame glasses, clear lenses
Representative image only.
Uvex by Honeywell Mfr # S3500 – Item # EW-86325-42
Available for sale in the US only
  • Lens features Ultra-dura™ coating for scratch and chemical resistance
  • Soft brow pad absorbs and diffuses energy of impact from flying debris
$15.50USD / Each
BUY MORE & SAVE
  • 1-9$15.50USD / Each
  • 10+$13.95USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorSilver/Navy
  • Lens ColorClear
  • Lens CoatingUltra-dura® (hard coating)
  • ComplianceANSI Z87+ (High Impact); military V0
  • Descriptionafety Glasses, Silver/Navy frame glasses, clear lenses
Key Features
  • Lens features Ultra-dura™ coating for scratch and chemical resistance
  • Soft brow pad absorbs and diffuses energy of impact from flying debris
More About this Item
Dual 9-base wrap-around lens provides uncompromised peripheral vision and side protection. Ventilation channels allow for air circulation around lens, minimizing fogging. Soft flexible nose fingers on nose bridge adjust and conform to any nasal profile and minimize slippage. Soft elastomer brow guard diffuses and deflects impact. The temples are ratcheting to allow for lens angle modification.
Compliance: ANSI Z87+ (High impact), military V0