Uvex by Honeywell S3810 Safety Goggles, Clear Lens, Indirect Vent
Representative image only.
Uvex by Honeywell Mfr # S3810 – Item # EW-86326-22
Available for sale in the US only
  • Uvextra™ AF lens coating minimizes fogging
  • Side loading rack has 4 drawes — each drawer holds 5 boxes
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorGray
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceANSI Z87.1-2010; CSA Z94.3
  • DescriptionSafety Goggles, Clear Lens, Indirect Vent
  • WarrantyLimited Lifetime
Canadian Standards AssociationLimited Lifetime Warranty
Key Features
  • Uvextra™ AF lens coating minimizes fogging
  • Side loading rack has 4 drawes — each drawer holds 5 boxes
  • Limited Lifetime Warranty
More About this Item
Lightweight, TPE body provides exceptional comfort and a snug fit for maximum protection. Fits over most prescription eyewear, and include Uvextra™ AF anti-fog lens coating that minimizes fogging.
Compliance ANSI Z87.1-2010 and CSA Z94.3 standards