Uvex by Honeywell S3850 Avatar Safety Glasses, Over-the-Glass, Black Frame, Clear, Antiscratch Hard Coat Lens; 10/BX
Representative image only.
Uvex by Honeywell Mfr # S3850 – Item # EW-86497-80
A custom fit each time with these highly adjustable wraparound glasses
  • Protect against impact from sun, wind dust and airborne debris Available in standard or Over-the Glass (OTG) glasses that go over your own corrective eyewear Lightweight 9-base wraparound frame design Ratchet lens has an angle-adjustable temple length for a more custom fit Soft, wire-core temples to customize to any facial profile Flexible nose pads to fit any nasal profile and reduce slippage Soft brow frame with indirect venting
  • Over-the-Glass Clear, Antiscratch Lens Black Frame
PRODUCT OPTIONS
$156.00USD / of 10
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorClear
  • Lens CoatingAntiscratch
  • Lens TypeOver-the-Glass
  • ComplianceANSI+ CSA Z94.3 (standard)
  • DescriptionSafety Glasses, Over-the-Glass, Black Frame, Clear, Antiscratch Hard Coat Lens; 10/BX
Key Features
  • Protect against impact from sun, wind dust and airborne debris Available in standard or Over-the Glass (OTG) glasses that go over your own corrective eyewear Lightweight 9-base wraparound frame design Ratchet lens has an angle-adjustable temple length for a more custom fit Soft, wire-core temples to customize to any facial profile Flexible nose pads to fit any nasal profile and reduce slippage Soft brow frame with indirect venting
  • Over-the-Glass Clear, Antiscratch Lens Black Frame
More About this Item
These high-performance glasses feature technology geared toward comfort, balance and protection from scratches, fog and more, while also providing the maximum field of vision. Polycarbonate lens features an antiscratch hard coating. Additional Hydroshield antifog or antireflective coating is also available.
Compliance: Certified to meet ANSI+ CSA Z94.3 (standard).