Uvex by Honeywell S3960C Safety Goggles, One Size, Gray Body/Clear Lens
Representative image only.
Uvex by Honeywell Mfr # S3960C – Item # EW-86326-00
Available for sale in the US only
  • Indirect ventilation and low profile design for impact and chemical splash protection
  • Uvextreme® coating for fog, scratch, and static resistance
  • Curved lens provides superior optics and peripheral vision
$23.75USD / Each
BUY MORE & SAVE
  • 1-9$23.75USD / Each
  • 10+$21.38USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorGray
  • Lens ColorClear
  • Lens CoatingUvextreme® (reduces fogging, scratches, and static)
  • ComplianceANSI Z87.1-2010; CSA Z94.3
  • DescriptionSafety Goggles, One Size, Gray Body/Clear Lens
Key Features
  • Indirect ventilation and low profile design for impact and chemical splash protection
  • Uvextreme® coating for fog, scratch, and static resistance
  • Curved lens provides superior optics and peripheral vision
More About this Item
Impact-resistant, low-profile design protects you from chemical splash—features indirect ventilation and a quick adjust headband. Uvextreme® anti-fog coating also resists scratches and static to ensure clear visibility. Polycarbonate lens gives maximum UV protection. Lens replacement snaps in quickly and easily.
Compliance: ANSI Z87.1-2010, CSA Z94.3