Uvex by Honeywell S3960HSI Stealth Safety Goggles, Gray Body, Fabric Headband, Clear, HydroShield Antifog Lens; 1/EA
Representative image only.
Uvex by Honeywell Mfr # S3960HSI – Item # EW-86497-92
Sleek style, comfort and protection all in one
  • Low-profile design and indirect venting system for chemical splash and impact protection Scratch-resistant and ideal for most indoor work HydroShield long-life antifog coating helps keep lenses clear Frame and lens are impact resistant
  • Gray Body Frame Fabric Headband
PRODUCT OPTIONS
$17.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorGray
  • Temple StyleFabric
  • Lens ColorClear
  • Lens CoatingAntifog
  • Lens TypeStandard
  • ComplianceANSI Z87.1-2015 and CSA Z94.3 standards
  • DescriptionSafety Goggles, Gray Body, Fabric Headband, Clear, HydroShield Antifog Lens; 1/EA
Key Features
  • Low-profile design and indirect venting system for chemical splash and impact protection Scratch-resistant and ideal for most indoor work HydroShield long-life antifog coating helps keep lenses clear Frame and lens are impact resistant
  • Gray Body Frame Fabric Headband
More About this Item
The soft elastomer construction of these goggles conforms to the face for a secure, comfortable and gap-free fit. Its elastomer seal stays soft, pliable and comfortable for the life of the frame. The goggles also feature a headband made of fabric or neoprene, a synthetic polymer that’s resistant to oil, heat and weathering.
There also is an Over-the-Glass (OTG) version of the goggle designed to fit comfortably over prescription eyewear. The safety goggles are clear and made of polycarbonate. They provide greater than 99.9% protection from harmful UV rays and come in other lens tints, too.
Compliance: ANSI Z87.1-2015 and CSA Z94.3 standards.