Uvex by Honeywell S9500 Turboshield Faceshield Headgear, Black
Representative image only.
Uvex by Honeywell Mfr # S9500 – Item # EW-86326-33
Comfortable, reliable headgear and face protection
  • Head-cushioning suspension cradle offers better support
  • Push-bottom visor attachment and release system are easy to use
  • Toric-shaped visor provides 50% more chin coverage
  • Easy to adjust large ergonomically designed control knobs
  • Hard hat adapter design enables visor to slide back
  • Dual-hinge adapter design increases clearance by as much as 4”
  • Compliance: ANSI Z87.1-2015 and CSA Z94.3
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFaceshield Headgear, Black
Key Features
  • Head-cushioning suspension cradle offers better support
  • Push-bottom visor attachment and release system are easy to use
  • Toric-shaped visor provides 50% more chin coverage
  • Easy to adjust large ergonomically designed control knobs
  • Hard hat adapter design enables visor to slide back
  • Dual-hinge adapter design increases clearance by as much as 4”
  • Compliance: ANSI Z87.1-2015 and CSA Z94.3
More About this Item
Whether it’s flying objects, debris, chemical splashes, or other hazards, these Turboshield faceshields protect the user’s entire face. Lightweight design is ideal for prolonged use. Order visors separately; they are available in clear, gray, or green UV/IR shade, as well as with a hard-coat and anti-fog protection. The headgear (86326-33) is sold separately.