Vaisala 19867HM Stainless Steel Grid Filter for Use with Hm46 Thermohygrometer Probe
Representative image only.
Vaisala Mfr # 19867HM – Item # EW-37801-20
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±1% RH; ±2% RH above 90% RH
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Temperature Accuracy±0.2°C
  • DescriptionStainless Steel Grid Filter for Use with Hm46 Thermohygrometer Probe
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Convenient parts and accessories for your thermohygrometer. Change capabilities, or keep a spare!