Vaisala HM70K3B1A1CB Thermohygrometer with Hmp76 12" L SS Probe and Alkaline Batteries
Representative image only.
Vaisala Mfr # HM70K3B1A1EB – Item # EW-37850-20
Dual probe input for multi-parameter measurement
  • Store 2700 data points—download later to your PC via RS-232 interface
  • Display measurements numerically or graphically
  • Three probe types available; tailor the meter for your application
  • Includes
  • MI70Link software with RS232 and analog output signal cables
  • Connection cable to HMD/W60/70 series
  • HMP76 12" long stainless steel humidity probe
  • Weather proof carrying case
  • Alkaline batteries
PRODUCT OPTIONS
$3,245.94USD / Each
sign in for your price.
$267.00 - $340.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Data LoggingYes
  • Battery4 AA (included)
  • DescriptionThermohygrometer with Hmp76 12\" L SS Probe and Alkaline Batteries
Includes
4 AA batteries.
Key Features
  • Store 2700 data points—download later to your PC via RS-232 interface
  • Display measurements numerically or graphically
  • Three probe types available; tailor the meter for your application
  • Includes
  • MI70Link software with RS232 and analog output signal cables
  • Connection cable to HMD/W60/70 series
  • HMP76 12" long stainless steel humidity probe
  • Weather proof carrying case
  • Alkaline batteries
More About this Item
Vaisala HM70 series meter measures temperature and humidity, plus calculates dew point, wet bulb, mixing ratio, absolute humidity, enthalpy, and water vapor pressure. It offers six different languages in display. Log three parameters into meter memory while displaying data in numerical or graphical fashion. View graphic trends of any parameter on the backlit LCD.
Customize your application with three humidity probes (sold separately). The general-purpose probe measures ambient conditions. Use the 12” stainless steel probe for air duct measurements. The extension cable probe includes a humidity sensor at the end of a 5-meter cable for high-temperature and hard-to-reach measurements. Order probes separately.