Value Plastics Barbed to Sanitary Adapter, ADCF Polypropylene; 1/4" ID to 1-1/2"
Representative image only.
Value Plastics Mfr # SFMX655-6005 – Item # EW-31513-55
No BSE documentation required!
  • Quickly connect tubing in your sanitary process with this 1-1/2" flange to 1/4" hose barb
PRODUCT OPTIONS
$28.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodEthylene oxide (ETO) or gamma
  • Connection TypeSanitary x Barb
  • Barb Size / Tubing Id (Port 2) (in)1/4
  • Sanitary Clamp Size (Port 1) (in)1-1/2
  • DescriptionBarbed to Sanitary Adapter, 1/4" ID to 1-1/2", ADCF-PP
Key Features
  • Quickly connect tubing in your sanitary process with this 1-1/2" flange to 1/4" hose barb
More About this Item
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8'' or 1/2'' hose barbs and 3/4'' mini or 1-1/2'' standard sanitary clamp connections.