Value Plastics Male Luer Thread 5/32" White Nylon 100/pk
Representative image only.
Value Plastics Mfr # MTLL025-1 – Item # EW-15973-17
Premium luer fittings for critical applications
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
$17.20USD / Pkg of 100
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
More About this Item
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.