Value Plastics Sanitary End Cap, ADCF Polypropylene; Mini 3/4"
Representative image only.
Value Plastics Mfr # SFMNC-6005 – Item # EW-31513-17
No BSE documentation required!
  • Sanitary end cap with handle fits on your mini 3/4" flange
PRODUCT OPTIONS
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodEthylene oxide (ETO) or gamma
  • Connection TypeSanitary x End cap
  • Sanitary Clamp Size (Port 1) (in)3/4
  • DescriptionSanitary End Cap, Mini 3/4", ADCF-PP
Key Features
  • Sanitary end cap with handle fits on your mini 3/4" flange
More About this Item
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8'' or 1/2'' hose barbs and 3/4'' mini or 1-1/2'' standard sanitary clamp connections.