Varipet 37895-0100 Repetitive Syringe Dispenser; 0 to 10.0 mL, 0.2 mL Divisions

Representative image only.
Varipet Mfr # 37895-0100 – Item # EW-07909-40
Self-filling pipetting aid is designed for speed, ease, and convenience
  • PTFE plunger and tip feature excellent chemical resistance
  • 6 month warranty

$305.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Max Volume (µL)10000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)10
  • Tip Volume (mL)10
  • Min Dispensing (mL)0
  • Max Dispensing (mL)10
  • Increments (mL)0.2
  • DescriptionRepetitive Syringe Dispenser; 0 to 10.0 mL, 0.2 mL Divisions
  • Warranty6 month
6 Month Warranty

Key Features

  • PTFE plunger and tip feature excellent chemical resistance
  • 6 month warranty

More About this Item

Simply turn the plunger knob to set your desired volume! These VARIPET dispensers are adjustable up to their total capacity. Once set, the plunger automatically returns to the same volume setting—providing you with a reproducibility of ±0.5% over the entire dispensing range.
Graduations on glass syringe barrel are easy to read. PTFE plunger provides smooth, self-lubricating, noncontaminating operation; ideal for use with most lab reagents and solvents except hydrofluoric acid. Molded PTFE tip. Choose from four volume ranges. Shpg wt 1 lb (0.5 kg).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat