Vector Laboratories Bloxall™ Dual-Enzyme Blocking Solution, 100 ml
Representative image only.
Vector Laboratories Mfr # SP-6000 – Item # EW-93951-71
Prepare the best samples possible
PRODUCT OPTIONS
$139.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)100
  • DescriptionDual-Enzyme Blocking Solution, 100 ml
More About this Item
Offering high sensitivity, low background, reliability, and reproducibility, these laboratory immunohistochemistry (IHC) slide preparation products accommodate a wide range of protocols.
BLOXALL™ Blocking Solution is a dual-enzyme blocking reagent that inactivates endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase in formalin-fixed, paraffin-embedded tissue sections, frozen tissue sections, and cell preparations. Simple one-step incubation takes only 10 minutes.