Vector Laboratories DuoLux Chemiluminescent/Fluorescent Substrate for Alkaline Phosphatase, 100 ml
Representative image only.
Vector Laboratories Mfr # SK-6605-100 – Item # EW-93952-09
Blots luminesce and fluoresce for easy visualization
  • Strong luminescent signal lasts up to 8 hours
  • Fluorescence is permanent and detectable for months
PRODUCT OPTIONS
$319.00USD / Each
Spectrum Chemical Agarose
Spectrum Chemical Agarose
$154.00 - $3,156.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorChemiluminescent, fluorescent
  • Volume (mL)100
  • ConjugateAlkaline Phosphatase
  • DescriptionChemiluminescent/Fluorescent Substrate for Alkaline Phosphatase, 100 ml
Key Features
  • Strong luminescent signal lasts up to 8 hours
  • Fluorescence is permanent and detectable for months
More About this Item
DuoLux substrate kits are available for both peroxidase and alkaline phosphatase reactions to produce a strong chemiluminescent and fluorescent signal. The substrate luminesces in the blue range, with a peak emission at 453 nm. Fluorescent excitation occurs over a range of wavelengths: 254 nm, 365 nm, with maximum excitation at 405 nm. Maximum fluorescent emission occurs at 453 nm.