Vector Laboratories EndTag™ 5' Nucleic Acid Labeling Kit
Representative image only.
Vector Laboratories Mfr # MB-9001 – Item # EW-93952-13
More efficient labeling of short oligonucleotides
  • Simple—get reliable, uniform labeling in approximately one hour
  • Diverse choice of labels: fluorophores, biotin, haptens, and affinity ligands (not included)
  • 1 year warranty
PRODUCT OPTIONS
$215.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description5' Nucleic Acid Labeling Kit
  • Warranty1 year
1 Year Warranty
Includes
T4 polynucleotide kinase, 10x reaction buffer, ATPγS, precipitant, and alkaline phosphatase.
Key Features
  • Simple—get reliable, uniform labeling in approximately one hour
  • Diverse choice of labels: fluorophores, biotin, haptens, and affinity ligands (not included)
  • 1 year warranty
More About this Item
EndTag DNA labeling systems are ideal for applications where an internal label might interfere with hybridization or sequence specific protein binding. Systems make labeling of short oligonucleotides more efficient and is an economical alternative to having labels inserted during synthesis. Both EndTag systems enable the covalent attachment of a variety of fluorescent dyes, haptens, or affinity tags to the respective ends of the nucleic acids using thiol-specific chemistry. Labels containing thiol-reactive groups can easily be incorporated. Note: Reactions require labeling reagents (Biotin maleimide, DNP maleimide, Fluorescein maleimide, Fucose maleimide, Texas Red® maleimide).
The 5’ EndTag Nucleic Acid Labeling System (93952-13) permits labeling of the 5’ ends of DNA, RNA, or unmodified oligonucleotides. It is ideal for PCR primers because it leaves the 3’ ends unmodified. Kit is designed to perform 10 labeling reactions of up to 0.6 nmols of 5’ ends per reaction.