Vector Laboratories H-3300 Citrate-Based Antigen Unmasking Solution, pH 6; 250 mL
Representative image only.
Vector Laboratories Mfr # H-3300-250 – Item # EW-93951-69
Prepare the best samples possible
  • 1 year warranty
PRODUCT OPTIONS
$222.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)250
  • DescriptionCitrate-Based Antigen Unmasking Solution, pH 6; 250 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Offering high sensitivity, low background, reliability, and reproducibility, these laboratory immunohistochemistry (IHC) slide preparation products accommodate a wide range of protocols.
Vector® Antigen Unmasking Solutions are highly effective at revealing antigens in formalin-fixed, paraffin embedded tissue sections when used in combination with a high temperature treatment. The solutions are available in two formulations: citrate-based with a pH 6.0 and Tris-based with a pH 9.0. Each bottle of concentrate is sufficient for preparation of 25 L of working solution.