Vector Laboratories ImmPACT AMEC Red, 120 ml
Representative image only.
Vector Laboratories Mfr # SK-4285 – Item # EW-93951-91
Successfully label multiple antigens in same tissue section
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
PRODUCT OPTIONS
$178.00USD / Each
Spectrum Chemical Agarose
Spectrum Chemical Agarose
$154.00 - $3,156.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorRed
  • Volume (mL)120
  • DescriptionAMEC Red, 120 ml
Key Features
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
More About this Item
These enzyme substrate kits for use with peroxidase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.
The substrates also come in a varying degree of sensitivities. The ImmPACT peroxidase substrates are about 2-4 times more sensitive than the original peroxidase substrate kits.
ImmPACT AMEC Red Substrate Kit produces a crisp, bright red reaction product that is 5 to 10 times more sensitive than conventional AEC substrates and can be used as a single label or as a second color for multiple antigen labeling.