Vector Laboratories ImmPACT Vector Red Substrate Kit, Red, 120 ml
Representative image only.
Vector Laboratories Mfr # SK-5105 – Item # EW-93952-03
Successfully label multiple antigens in same tissue section
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
  • 1 year warranty
PRODUCT OPTIONS
$211.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorRed
  • Volume (mL)120
  • DescriptionVector Red Substrate Kit, Red, 120 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
  • 1 year warranty
More About this Item
These enzyme substrate kits for use with alkaline phosphatase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.
Vector Red and ImmPACT Red Substrate Kits produce red, heat stable reaction products that are highly fluorescent and non-fading. Because of their stability under ISH conditions, both chromogens can be used in IHC/ISH double labeling applications.