Vector Laboratories ImmPRINT™ Solvent-Resistant Histology Pen, 5/pk
Representative image only.
Vector Laboratories Mfr # H-6100 – Item # EW-93951-67
Prepare the best samples possible
  • Marking pens ideal for aqueous and solvent environments
  • 1 year warranty
PRODUCT OPTIONS
$125.00USD / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSolvent-Resistant Histology Pen, 5/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Marking pens ideal for aqueous and solvent environments
  • 1 year warranty
More About this Item
Offering high sensitivity, low background, reliability, and reproducibility, these laboratory immunohistochemistry (IHC) slide preparation products accommodate a wide range of protocols.
ImmPrint™ Histology Pen is a solvent-resistant permanent marking pen designed for writing on frosted glass microscope slides, tissue cassettes, and most hard surfaces. The pen’s smooth writing tip resists drying out, while the high density, fast-drying black ink is resistant to most organic solvents encountered in histological applications.
For sale in U.S. only!