Vector Laboratories S-2012-50 Normal Horse Serum, 2.5% solution (v/v); 50 mL

Representative image only.
Vector Laboratories Mfr # S-2012-50 – Item # EW-93951-76
Ideal for use in blocking nonspecific binding
  • Supplied undiluted with 0.08% sodium azide as a preservative
  • Applications include immunohistochemistry, immunocytochemistry, and immunofluorescence
  • 1 year warranty

PRODUCT OPTIONS

$140.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)50
  • DescriptionNormal Horse Serum, 2.5% solution (v/v); 50 mL
  • Warranty1 year
1 Year Warranty

Key Features

  • Supplied undiluted with 0.08% sodium azide as a preservative
  • Applications include immunohistochemistry, immunocytochemistry, and immunofluorescence
  • 1 year warranty

More About this Item

Choose from a variety of sera, which when diluted can be used for blocking nonspecific binding or as an antibody diluent. The sera are pooled samples collected from healthy adult goats, horses, chickens, swine, rabbits, cats, and dogs. Each one is heat-treated and centrifuged to remove precipitates and then filtered. The sera are tested with appropriate antibody to check for possible cross-reactivity.

Cat and dog sera can be added to secondary antibody diluent to adsorb potential cross-reactivity with each animal’s tissue.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat