Vector Laboratories SK-5400 BCIP/NBT Substrate Kit, Indigo, 200 ml
Representative image only.
Vector Laboratories Mfr # SK-5400 – Item # EW-93952-06
Successfully label multiple antigens in same tissue section
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
PRODUCT OPTIONS
$182.00USD / Each
Spectrum Chemical Agarose
Spectrum Chemical Agarose
$154.00 - $3,156.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorIndigo
  • Volume (mL)200
  • DescriptionBCIP/NBT Substrate Kit, Indigo, 200 ml
Key Features
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
More About this Item
These enzyme substrate kits for use with alkaline phosphatase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.
The substrates also come in a varying degree of sensitivities. The alkaline phosphatase substrate BCIP/NBT can achieve increased sensitivities with longer incubation times. BCIP/NBT Substrate Kit forms an indigo, heat stable reaction product that can be dehydrated, cleared, and permanently mounted. It can be used singly or in combination with other alkaline phosphatase or peroxidase substrates for multiple label applications.