Vector Laboratories SK-5900 p-Nitrophenylphosphate Concentrate, Yellow, 18 ml
Representative image only.
Vector Laboratories Mfr # SK-5900 – Item # EW-93952-07
Successfully label multiple antigens in same tissue section
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ColorYellow
  • Volume (mL)360
  • Descriptionp-Nitrophenylphosphate Concentrate, Yellow, 18 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
  • 1 year warranty
More About this Item
These enzyme substrate kits for use with alkaline phosphatase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.
p-Nitrophenylphosphate Substrate Kit is ideal for ELISA using alkaline phosphatase enzyme detection systems. The bright yellow color of the soluble reaction product is measured at 405-420 nm, providing an excellent reagent for microtitration plate readers.