Vector Laboratories Tris-based Antigen Unmasking Solution, pH 9; 250 mL
Representative image only.
Vector Laboratories Mfr # H-3301-250 – Item # EW-93951-70
Prepare the best samples possible
PRODUCT OPTIONS
$173.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)250
  • DescriptionTris-based Antigen Unmasking Solution, pH 9, 25 L
More About this Item
Offering high sensitivity, low background, reliability, and reproducibility, these laboratory immunohistochemistry (IHC) slide preparation products accommodate a wide range of protocols.
Vector® Antigen Unmasking Solutions are highly effective at revealing antigens in formalin-fixed, paraffin embedded tissue sections when used in combination with a high temperature treatment. The solutions are available in two formulations: citrate-based with a pH 6.0 and Tris-based with a pH 9.0. Each bottle of concentrate is sufficient for preparation of 25 L of working solution.