Vector Laboratories VectaCell Trolox Antifade Reagent for Live Cell Imaging, 2 ml
Representative image only.
Vector Laboratories Mfr # CB-1000-2 – Item # EW-93952-18
Enhances contrast of fixed and live cell images
  • 1 year warranty
$143.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorColorless
  • Volume (mL)2
  • DescriptionTrolox Antifade Reagent for Live Cell Imaging, 2 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The VectaCell stains are ideal for conventional or super-resolution fluorescence microscopy, immunocytochemistry, in situ hybridization or neurobiology applications on fixed or live cells. Several stains are available.
VectaCell Trolox is an anti-fading additive that reduces photobleaching and blinking during live cell imaging. It is a water-soluble and cell-permeable analog of vitamin E, that prevents formation of different reactive oxygen species, such as singlet oxygen, superoxide anion, or hydrogen peroxide. Trolox has been shown to have a cytoprotective effect and low cytotoxicity for different cell lines.