Vector Laboratories VECTASHIELD® Antifade Mounting Medium with PI, 10 ml
Representative image only.
Vector Laboratories Mfr # H-1300-10 – Item # EW-93952-25
Prepare the best samples possible
  • Mounting media for fluorescence that are stable and control photobleaching
  • 1 year warranty
PRODUCT OPTIONS
$254.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)10
  • DescriptionAntifade Mounting Medium with PI, 10 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Mounting media for fluorescence that are stable and control photobleaching
  • 1 year warranty
More About this Item
Offering high sensitivity, low background, reliability, and reproducibility, these laboratory immunohistochemistry (IHC) slide preparation products accommodate a wide range of protocols.
VECTASHIELD® Mounting Medias feature a stable formula for preserving fluorescence, while also preventing rapid photobleaching of fluorescent proteins and dyes. Nonhardening formulations remain as a liquid on the slide and can be stored without sealing; the coverslip can be sealed around the perimeter with nail polish or plastic sealant. HardSet™ formulations harden after coverslipping; the coverslip becomes immobilized after 15 minutes at room temperature offering optimal antifade ability and refractive index.