Vector Laboratories WestVision Block and Diluent for Western Blots, 500 ml
Representative image only.
Vector Laboratories Mfr # SP-7000-500 – Item # EW-93952-10
Improve your signal intensity without increasing background
  • Save time with ready-to-use format
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)500
  • DescriptionBlock and Diluent for Western Blots, 500 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Save time with ready-to-use format
  • 1 year warranty
More About this Item
WestVision™ Block and Diluent is a ready-to-use Tris buffer-based blocking solution intended for Western or dot blot applications. This solution is a proprietary formulation containing Tween 20 that improves signal intensity and resolution without increasing background. To achieve the best results, use WestVision™ Block and Diluent for all blocking and diluting steps including primary antibody and secondary antibody detection reagents. WestVision™ Block and Diluent is compatible with both alkaline phosphatase- and peroxidase- based detections systems, as well as with both chemiluminescence and chromogenic development.
A method to improve signal intensity and resolution without increasing background is to use the WestVision Block and Diluent. This ready-to-use Tris buffer-based blocking solution containing Tween 20, is intended specifically for Western blot applications. It is ideal for all blocking and diluting steps, including primary antibody and secondary antibody detection reagents. The solution is compatible with both alkaline phosphatase- and peroxidase-based detections systems, as well as with both chemiluminescence and chromogenic development.