Venture Tech CAP24 End cap, PFA, for 1, 1-1/2" sanitary clamp
Representative image only.
Venture Tech Mfr # CAP24 – Item # EW-31530-35
Ideal for extremely corrosive materials—PFA will not rouge or pit
  • Inert, nonstick surfaces—perfect for fragile and metal-sensitive proteins
  • Employ regular steam-in-place cycles without worry
  • Fully compatible with other Tri-Clamp® connections
  • Protective overmolding reinforces clamp connection sites
  • Full static vacuum capability at 300°F (149°C)
PRODUCT OPTIONS
$121.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Sterilization MethodAutoclave only
  • Connection TypeSanitary
  • Sanitary Clamp Size (Port 1) (in)1-1/2
  • Max Temperature (° F)180 at 14.7 psig
  • Max Temperature (° C)82 at 1 bar
  • Max Operating Pressure (PSI)100 psi in ambient conditions; 60 psi at 180°F (82°C); 60 psi at 35°F (2°C)
  • Max Vacuum (in Hg)32
  • Max Vacuum (mm Hg)812
  • DescriptionEnd cap, PFA, for 1, 1-1/2" sanitary clamp
Key Features
  • Inert, nonstick surfaces—perfect for fragile and metal-sensitive proteins
  • Employ regular steam-in-place cycles without worry
  • Fully compatible with other Tri-Clamp® connections
  • Protective overmolding reinforces clamp connection sites
  • Full static vacuum capability at 300°F (149°C)
More About this Item
Sanitary applications that use highly corrosive chemicals require fittings that can endure the punishment. These food and pharmaceutical quality PFA fittings are easily cleaned so you will minimize downtime, cross contamination, and yield loss. Autoclavable