Vici Metronics LM44032 Gc801:So2 Toxic Gas Monitor/Alarm
Representative image only.
Vici Metronics Mfr # LM44032 – Item # EW-10110-01
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Battery Type1 x 9V battery
  • DescriptionGc801:So2 Toxic Gas Monitor/Alarm
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
A complete range of small, portable monitors/alarms for the detection of O2 and toxic gases, such as CO, H2S, SO2, NOx, & NO2. Ideally suited as a personal monitor for field use, each unit is dedicated for the continuous measurement and alarm of a particular gas.
FEATURES
  • Pocket-size, light weight
  • Continuously monitors and displays concentrations on LCD
  • Audible and visual alarms
  • Operates on a single 9 volt battery
  • Wide temperature operating range
  • Uses GCI electrochemical sensors
  • Large LCD display