This item is now discontinued. We recommend the following as suggested replacements: EW-98766-58
Vision USA 6971 Handheld LED Illuminated Magnifier, 2" Dia Viewing Area; 7x
Representative image only.
Vision USA Mfr # 127078 – Item # EW-03889-80
High quality LED illumination
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification7x
  • Viewing Area2\" (50.8 mm) diameter
  • LensSingle
  • Mounting TypeHandheld
  • Light SourceLED
  • BatteryAA Battery (2)
  • DescriptionHandheld LED Illuminated Magnifier, 2\" Dia Viewing Area; 7x
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The large viewing area and brighter light provides greater detail and less eye-strain during inspections.
Handheld models feature a compact, pocket-sized design—ideal for field use. Handheld magnifiers require three AAA batteries (sold separately).