VistaLab 1066 Fixed Volume Pipette, 2.0 mL, Red; Each

Representative image only.
VistaLab Mfr # 1066 – Item # EW-06199-17
Ideal for tasks requiring high accuracy and large volumes
  • Convenient, color-coded, fixed-volume pipets
  • High accuracy and precision
  • Simple single-stroke procedure
  • Limited Lifetime Warranty

PRODUCT OPTIONS

$367.00USD / Each

$89.50 - $162.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought With…

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Max Volume (µL)2000
  • Max Volume (mL)2
  • Max Dispensing (mL)2
  • Accuracy (%)±0.75%
  • Precision (%)±0.50%
  • Overall Length (in)8.9
  • Overall Length (mm)226
  • Color CodeRed
  • Compatible withVistaLab 5 mL tips
  • WarrantyLimited Lifetime
Limited Lifetime Warranty

Key Features

  • Convenient, color-coded, fixed-volume pipets
  • High accuracy and precision
  • Simple single-stroke procedure
  • Limited Lifetime Warranty

More About this Item

Reconstitute your reagents, controls, or dilutions using these precise MLA Macro Pipettors. Calibrated "to deliver" instruments—they have a fixed stroke and consistently deliver the stated or calibrated volume when the plunger is fully depressed. Each pipet can be adjusted to deliver above or below its stated volume within ±10%. Calibration is accomplished readily by inserting a small key into the plunger and turning the key.
These pipettes are color-coded for easy identification and come with the stated volume engraved onto the piston assembly. Constructed of aluminum alloy and stainless steel, these pipettors withstand years of use and provide exceptional performance for a wide range of fluids — including dense or viscous samples.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat