VistaLab Protectainer™ Pipette Tips, Non-Sterile, Bulk Pack, 250 µL; 1000/Pk
Representative image only.
VistaLab Mfr # 4025 – Item # EW-06199-79
Maximize performance and benefits of your pipette
  • Tips made of 100% pure virgin polypropylene
PRODUCT OPTIONS
$65.00USD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)250
  • Min Volume (µL)1
  • Overall Length (in)2.16
  • Overall Length (mm)54.8
  • Wetted MatterialPolypropylene
  • Package OfBulk; 1000/Pk
  • Compatible withOvation ESC, E8, E12, QS, M, F1, F2 & All MLA model Pipettors
  • DescriptionPipette Tips, Non-Sterile, Bulk Pack, 250 µL; 1000/Pk
Key Features
  • Tips made of 100% pure virgin polypropylene
More About this Item
Protectainer™ polypropylene (PP) tips come bulk packaged in a plastic storage container with snap-on lid, which may be recycled. Tips are compatible with VistaLab™ Ovation and MLA model pipettors.