Waring 7010HS blender 2-speed, 1-liter, SS, 120v
Representative image only.
Waring Mfr # 7010HS – Item # EW-04245-42
  • 1 year warranty
PRODUCT OPTIONS
$1,084.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)1
  • Min RPM18000
  • Max RPM22000
  • Motor TypeHeavy-duty
  • Power (VAC)120
  • Power (amps)7
  • Power (Hz)50/60
  • Container MaterialStainless Steel
  • Timer (minutes)3
  • ColorWhite
  • Descriptionblender 2-speed, 1-liter, SS, 120v
  • Warranty1 Year
1 Year WarrantyUL Listed.
Key Features
  • 1 year warranty
More About this Item
Two-speed blenders with timer offer a mechanical 3-minute timer that allows automatic shutoff and a push-button speed control for 18,000 and 22,000 rpm operation.
Variable-speed controller, available separately, lets you adjust speed from 1,000 rpm to the maximum speed.